siRNA Details
| Gene Symbol: Vps8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001285893.1 | NP_001272822.1 | 34, 35 | 3302 | ||
| GenBank mRNA | AK029007.1 | - | 18, 19 | 1519 | ||
| AK031205.1 | - | 11, 12 | 1344 | |||
| AK141678.1 | - | 34, 35 | 3225 | |||
| AK149336.1 | - | 15, 16 | 1381 | |||
| BC023914.1 | - | 20, 21 | 1693 | 5323126 | ||
| BC032214.1 | - | 14, 15 | 1271 | 5323693 | ||
| BC055323.1 | - | 34, 35 | 3219 | 5706288 | ||

Vps8
Genomic Map