siRNA Details
| Gene Symbol: Taf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027748.3 | NP_082024.2 | 1 | 226 | ||
| GenBank mRNA | AJ292189.1 | - | 1 | 17 | ||
| AK154372.1 | - | 1 | 235 | |||
| BB621106.1 | - | 1 | 227 | |||
| BC039989.1 | - | 1 | 182 | |||
| BC089030.1 | - | 1 | 65 | |||
| BC094546.1 | - | 1 | 183 | |||
| BC137617.1 | - | 1 | 167 | 8860636 | ||
| BC137618.1 | - | 1 | 167 | 8860637 | ||
| Gene Symbol: C630004M23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK049866.1 | - | 1 | 545 | ||

Taf3
Genomic Map