siRNA Details
| Gene Symbol: Swap70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009302.3 | NP_033328.3 | 2 | 313 | ||
| GenBank mRNA | AF053974.1 | - | 2 | 205 | ||
| AK019882.1 | - | 2 | 256 | |||
| AK041505.1 | - | 2 | 261 | |||
| AK045295.1 | - | 2 | 285 | |||
| AK146773.1 | - | 2 | 305 | |||
| AK172990.1 | - | 2 | 250 | |||
| BC065136.1 | - | 2 | 255 | 30546814 | ||
| BY118458.1 | - | 2 | 314 | |||

Swap70
Genomic Map