siRNA Details
| Gene Symbol: Rgag1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001040434.2 | NP_001035524.2 | 1 | 2941 | ||
| GenBank mRNA | AK220382.1 | - | 1 | 2311 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rgag1| Gene Symbol: Rgag1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001040434.2 | NP_001035524.2 | 1 | 2941 | ||
| GenBank mRNA | AK220382.1 | - | 1 | 2311 | ||