siRNA Details
| Gene Symbol: Ttc28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001267622.1 | NP_001254551.1 | 19, 20 | 5631 | ||
| GenBank mRNA | AK031458.1 | - | 2, 3 | 225 | ||
| AK081382.1 | - | 2, 3 | 103 | |||
| AK122427.1 | - | 13, 14 | 3194 | |||
| BC046779.1 | - | 13, 14 | 3295 | 5719497 | ||
| BC060735.1 | - | 12, 13 | 2291 | |||

Ttc28
Genomic Map