siRNA Details
Gene Symbol: Lcorl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163073.1 | NP_001156545.1 | 2 | 240 | ||
NM_172153.3 | NP_742165.1 | 2 | 253 | |||
NM_178142.5 | NP_835278.2 | 2 | 240 | |||
GenBank mRNA | AB076078.1 | - | 2 | 252 | ||
AK041987.1 | - | 2 | 199 | |||
AK042567.1 | - | 2 | 254 | |||
AK043498.1 | - | 2 | 198 | |||
AK043845.1 | - | 2 | 208 | |||
AK083689.1 | - | 2 | 202 | |||
AK154398.1 | - | 2 | 201 | |||
AK155421.1 | - | 2 | 201 | |||
BC066151.1 | - | 2 | 170 | |||
BC138563.1 | - | 2 | 240 | 8861583 |
