siRNA Details
| Gene Symbol: Synj2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113351.1 | NP_001106822.1 | 2, 3 | 565 | ||
| NM_001113352.2 | NP_001106823.1 | 3, 4 | 654 | |||
| NM_001113353.2 | NP_001106824.1 | 3, 4 | 654 | |||
| NM_011523.2 | NP_035653.2 | 2, 3 | 565 | |||
| GenBank mRNA | AF041857.1 | - | 3, 4 | 655 | ||
| AF041862.1 | - | 2, 3 | 510 | |||
| AK148053.1 | - | 3, 4 | 393 | |||
| BC058749.1 | - | 3, 4 | 531 | 6390688 | ||
| BC060214.1 | - | 2, 3 | 411 | 6838657 | ||

Synj2
Genomic Map