siRNA Details
| Gene Symbol: Dennd2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172477.4 | NP_766065.1 | 2 | 483 | ||
| GenBank mRNA | AK047138.1 | - | 3 | 471 | ||
| AK048041.1 | - | 2 | 467 | |||
| AK081176.1 | - | 2 | 484 | |||
| AK160784.1 | - | 2 | 245 | |||
| AK165202.1 | - | 2 | 339 | |||
| AK167298.1 | - | 2 | 337 | |||
| BC056367.1 | - | 2 | 362 | |||
| BC057310.1 | - | 2 | 362 | |||
| BC145599.1 | - | 1 | 165 | 9054165 | ||
| BC150799.1 | - | 1 | 165 | 9087710 | ||

Dennd2a
Genomic Map