siRNA Details
| Gene Symbol: Metrnl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144797.3 | NP_659046.1 | 2 | 641 | ||
| GenBank mRNA | AK034855.1 | - | 2 | 642 | ||
| AK170800.1 | - | 2 | 643 | |||
| AK171200.1 | - | 2 | 387 | |||
| BC019776.1 | - | 2 | 596 | 3672224 | ||
| BC024445.1 | - | 2 | 656 | 4974030 | ||
| BC024497.1 | - | 644 | 4982563 | |||
| BC026646.1 | - | 2 | 635 | 4987595 | ||

Metrnl
Genomic Map