siRNA Details
| Gene Symbol: Zfp719 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172482.1 | NP_766070.1 | 4 | 1916 | ||
| GenBank mRNA | AK033195.1 | - | 1 | 1295 | ||
| AK083143.1 | - | 4 | 1857 | |||
| BC035197.1 | - | 1 | 876 | |||
| BC081548.1 | - | 4 | 1946 | |||

Zfp719
Genomic Map