siRNA Details
| Gene Symbol: Zfp719 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172482.1 | NP_766070.1 | 3, 4 | 528 | ||
| GenBank mRNA | AK044684.1 | - | 3, 4 | 505 | ||
| AK083143.1 | - | 3, 4 | 469 | |||
| BC081548.1 | - | 3, 4 | 491 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp719| Gene Symbol: Zfp719 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172482.1 | NP_766070.1 | 3, 4 | 528 | ||
| GenBank mRNA | AK044684.1 | - | 3, 4 | 505 | ||
| AK083143.1 | - | 3, 4 | 469 | |||
| BC081548.1 | - | 3, 4 | 491 | |||