siRNA Details
| Gene Symbol: Irgc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199013.2 | NP_950178.2 | 3 | 1707 | ||
| GenBank mRNA | AK089224.1 | - | 3 | 1710 | ||
| AK133005.1 | - | 2 | 1609 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Irgc1| Gene Symbol: Irgc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199013.2 | NP_950178.2 | 3 | 1707 | ||
| GenBank mRNA | AK089224.1 | - | 3 | 1710 | ||
| AK133005.1 | - | 2 | 1609 | |||