siRNA Details
| Gene Symbol: Shank2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081370.2 | NP_001074839.2 | 11 | 4046 | ||
| NM_001113373.2 | NP_001106844.2 | 16 | 4492 | |||
| GenBank mRNA | AB099695.1 | - | 16 | 4131 | ||
| AK220363.1 | - | 9 | 3061 | |||
| CF540072.1 | - | 2 | 435 | |||

Shank2
Genomic Map