siRNA Details
| Gene Symbol: Shank2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081370.2 | NP_001074839.2 | 1 | 694 | ||
| NM_001113373.2 | NP_001106844.2 | 6 | 1140 | |||
| GenBank mRNA | AB099695.1 | - | 6 | 767 | ||
| AK139360.1 | - | 6 | 1141 | |||
| CJ109874.1 | - | 1 | 695 | |||

Shank2
Genomic Map