siRNA Details
| Gene Symbol: Gm12169 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163357.1 | NP_001156829.1 | 7 | 761 | ||
| GenBank mRNA | BG066735.2 | - | 1 | 262 | ||
| C80168.1 | - | 1 | 259 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm12169| Gene Symbol: Gm12169 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163357.1 | NP_001156829.1 | 7 | 761 | ||
| GenBank mRNA | BG066735.2 | - | 1 | 262 | ||
| C80168.1 | - | 1 | 259 | |||