siRNA Details
Gene Symbol: Gm12169 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163357.1 | NP_001156829.1 | 7 | 788 | ||
GenBank mRNA | BG066735.2 | - | 1 | 235 | ||
C80168.1 | - | 1 | 232 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm12169 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163357.1 | NP_001156829.1 | 7 | 788 | ||
GenBank mRNA | BG066735.2 | - | 1 | 235 | ||
C80168.1 | - | 1 | 232 |