siRNA Details
| Gene Symbol: Themis | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB509340.1 | - | 3 | 688 | ||
| AK040594.1 | - | 3 | 867 | |||
| AK041386.1 | - | 3 | 904 | |||
| AK041562.1 | - | 3 | 793 | |||
| AK041622.1 | - | 3 | 867 | |||
| AK088136.1 | - | 3 | 869 | |||
| AK089077.1 | - | 3 | 869 | |||
| AK138701.1 | - | 3 | 867 | |||
| AK169558.1 | - | 4 | 799 | |||
| BC125486.1 | - | 3 | 750 | 40130001 | ||
| BC125488.1 | - | 3 | 750 | 40130003 | ||

Themis
Genomic Map