siRNA Details
| Gene Symbol: Ncoa7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001111267.2 | NP_001104737.1 | 4 | 584 | ||
| NM_172495.5 | NP_766083.3 | 14 | 2935 | |||
| GenBank mRNA | AK033503.1 | - | 4 | 498 | ||
| AK042369.1 | - | 4 | 554 | |||
| AK128935.1 | - | 13 | 2758 | |||
| AK165098.1 | - | 4 | 553 | |||
| AK171679.1 | - | 4 | 502 | |||
| BC042791.1 | - | 1015 | ||||
| BC049997.1 | - | 4 | 534 | |||
| BC058572.1 | - | 4 | 506 | |||
| BC076623.1 | - | 12 | 2541 | |||
| BC080670.1 | - | 508 | ||||
| BC080742.1 | - | 3 | 1690 | |||
| BC141150.1 | - | 13 | 2636 | 9055762 | ||
| BC145459.1 | - | 12 | 2521 | |||
| EF602028.1 | - | 4 | 584 | |||

Ncoa7
Genomic Map