siRNA Details
| Gene Symbol: Kcnj14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145963.2 | NP_666075.1 | 2 | 358 | ||
| GenBank mRNA | AK030711.1 | - | 2 | 368 | ||
| AK044725.1 | - | 2 | 339 | |||
| AK080732.1 | - | 2 | 347 | |||
| BC029692.1 | - | 2 | 298 | 5363132 | ||
| BC031753.1 | - | 2 | 358 | 4503027 | ||

Kcnj14
Genomic Map