siRNA Details
| Gene Symbol: Zfyve26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008550.1 | NP_001008550.1 | 28 | 5576 | ||
| GenBank mRNA | AK042386.1 | - | 2 | 919 | ||
| AK051350.1 | - | 1 | 571 | |||
| AK169541.1 | - | 2 | 917 | |||
| AK220333.1 | - | 28 | 5547 | |||
| BC047061.1 | - | 8 | 1142 | |||
| BC048379.1 | - | 8 | 1153 | |||
| BC141359.1 | - | 28 | 5559 | 9055971 | ||

Zfyve26
Genomic Map