siRNA Details
| Gene Symbol: Tmem60 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177601.3 | NP_808269.1 | 2 | 266 | ||
| GenBank mRNA | AK041036.1 | - | 2 | 238 | ||
| BC027828.1 | - | 1 | 43 | 5249625 | ||
| CN834017.1 | - | 2 | 532 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem60| Gene Symbol: Tmem60 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177601.3 | NP_808269.1 | 2 | 266 | ||
| GenBank mRNA | AK041036.1 | - | 2 | 238 | ||
| BC027828.1 | - | 1 | 43 | 5249625 | ||
| CN834017.1 | - | 2 | 532 | |||