siRNA Details
| Gene Symbol: Rln3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173184.1 | NP_775276.1 | 1 | 154 | ||
| GenBank mRNA | AB076565.1 | - | 1 | 154 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rln3| Gene Symbol: Rln3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173184.1 | NP_775276.1 | 1 | 154 | ||
| GenBank mRNA | AB076565.1 | - | 1 | 154 | ||