siRNA Details
| Gene Symbol: Proser1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173382.1 | NP_775558.2 | 11 | 2665 | ||
| GenBank mRNA | AK034174.1 | - | 11 | 2823 | ||
| AK038404.1 | - | 11 | 2665 | |||
| BC025559.1 | - | 1 | 782 | |||
| BC051453.1 | - | 1 | 1433 | |||
| BC057544.1 | - | 3 | 1575 | |||
| BC079616.1 | - | 1293 | ||||
| BC086674.1 | - | 11 | 2222 | 30653602 | ||

Proser1
Genomic Map