siRNA Details
| Gene Symbol: Proser1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173382.1 | NP_775558.2 | 4 | 668 | ||
| GenBank mRNA | AK034174.1 | - | 4 | 819 | ||
| AK038404.1 | - | 4 | 668 | |||
| AK087909.1 | - | 4 | 689 | |||
| AK161440.1 | - | 3 | 487 | |||
| AK199697.1 | - | 4 | 233 | |||
| AK207125.1 | - | 4 | 233 | |||
| BC086674.1 | - | 4 | 225 | 30653602 | ||

Proser1
Genomic Map