siRNA Details
| Gene Symbol: Proser1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173382.1 | NP_775558.2 | 5 | 770 | ||
| GenBank mRNA | AK034174.1 | - | 5 | 921 | ||
| AK038404.1 | - | 5 | 770 | |||
| AK087909.1 | - | 5 | 791 | |||
| AK161440.1 | - | 4 | 589 | |||
| AK207125.1 | - | 5 | 335 | |||
| BC086674.1 | - | 5 | 327 | 30653602 | ||

Proser1
Genomic Map