siRNA Details
| Gene Symbol: Klhl32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033531.3 | NP_001028703.2 | 5 | 804 | ||
| NM_001163020.1 | NP_001156492.1 | 6 | 903 | |||
| GenBank mRNA | AK122569.1 | - | 526 | |||
| AK134944.1 | - | 5 | 776 | |||
| BC141239.1 | - | 5 | 508 | 9055851 | ||
| BC145566.1 | - | 4 | 404 | |||
| CA556185.2 | - | 5 | 495 | |||

Klhl32
Genomic Map