siRNA Details
| Gene Symbol: Mtg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199301.2 | NP_955005.2 | 10, 11 | 965 | ||
| GenBank mRNA | AK144453.1 | - | 10, 11 | 890 | ||
| AK144625.1 | - | 10, 11 | 923 | |||
| BC027306.1 | - | 10, 11 | 874 | 4019717 | ||

Mtg1
Genomic Map