siRNA Details
| Gene Symbol: Iffo2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001205173.1 | NP_001192102.1 | 5 | 1295 | ||
| NM_183148.3 | NP_898971.1 | 8 | 915 | |||
| GenBank mRNA | BC100381.1 | - | 8 | 915 | ||
| BE860309.1 | - | 3 | 284 | |||
| CN718972.1 | - | 2 | 259 | |||
| EH105040.1 | - | 2 | 231 | |||

Iffo2
Genomic Map