siRNA Details
| Gene Symbol: Slc45a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177628.2 | NP_001171099.1 | 3 | 846 | ||
| NM_145977.2 | NP_666089.1 | 3 | 927 | |||
| GenBank mRNA | AK035428.1 | - | 3 | 796 | ||
| AK162123.1 | - | 3 | 774 | |||
| BC031381.1 | - | 3 | 927 | 5050610 | ||
| BC034084.1 | - | 1 | 176 | |||

Slc45a3
Genomic Map