siRNA Details
| Gene Symbol: Pdlim2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001253736.1 | NP_001240665.1 | 5 | 474 | ||
| NM_145978.2 | NP_666090.1 | 5 | 401 | |||
| GenBank mRNA | AK137991.1 | - | 4 | 457 | ||
| BC024556.1 | - | 5 | 401 | 4990983 | ||
| DV646822.1 | - | 5 | 374 | |||

Pdlim2
Genomic Map