siRNA Details
| Gene Symbol: T2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001161832.1 | NP_001155304.1 | 9, 10 | 1026 | ||
| GenBank mRNA | AF072546.1 | - | 6, 7 | 618 | ||
| BC119261.1 | - | 3, 4 | 384 | 8734010 | ||
| BC119287.1 | - | 3, 4 | 384 | 8734036 | ||

T2
Genomic Map