siRNA Details
| Gene Symbol: Dstyk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172516.4 | NP_766104.2 | 3 | 953 | ||
| GenBank mRNA | AK020880.1 | - | 3 | 812 | ||
| AK083696.1 | - | 3 | 851 | |||
| AK132119.1 | - | 3 | 828 | |||
| AK145449.1 | - | 3 | 798 | |||
| AK172954.1 | - | 3 | 800 | |||
| AK186755.1 | - | 1 | 23 | |||
| AK205064.1 | - | 1 | 125 | |||
| AY208852.1 | - | 3 | 800 | |||
| AY429676.1 | - | 3 | 839 | |||
| BC130226.1 | - | 3 | 829 | 40141770 | ||

Dstyk
Genomic Map