siRNA Details
| Gene Symbol: Tagln | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011526.5 | NP_035656.1 | 2 | 346 | ||
| GenBank mRNA | AK002880.1 | - | 2 | 185 | ||
| AK077235.1 | - | 2 | 187 | |||
| BC003795.1 | - | 2 | 175 | 3600413 | ||
| L41154.1 | - | 2 | 185 | |||
| U36588.1 | - | 2 | 185 | |||
| Gene Symbol: Pcsk7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK028131.1 | - | 1538 | |||

Tagln
Genomic Map