siRNA Details
| Gene Symbol: Szrd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025608.2 | NP_001020779.1 | 3 | 309 | ||
| NM_001277195.1 | NP_001264124.1 | 3 | 306 | |||
| GenBank mRNA | AK027934.1 | - | 2 | 222 | ||
| AK145912.1 | - | 2 | 229 | |||
| AK171420.1 | - | 2 | 219 | |||
| AK171702.1 | - | 2 | 234 | |||
| BB871722.1 | - | 2 | 236 | |||
| BC066992.1 | - | 3 | 257 | 5685985 | ||
| CV784607.1 | - | 3 | 248 | |||

Szrd1
Genomic Map