siRNA Details
| Gene Symbol: Tarbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009319.3 | NP_033345.2 | 2 | 487 | ||
| GenBank mRNA | AK143188.1 | - | 2 | 488 | ||
| AV454829.1 | - | 2 | 175 | |||
| AV582078.1 | - | 2 | 345 | |||
| BC002028.1 | - | 2 | 200 | 3489376 | ||
| BI737851.1 | - | 2 | 434 | |||
| BQ948575.1 | - | 2 | 228 | |||
| U79962.1 | - | 2 | 220 | |||

Tarbp2
Genomic Map