siRNA Details
| Gene Symbol: Duoxa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK042259.1 | - | 2 | 316 | ||
| BC019755.1 | - | 8 | 781 | 3964798 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Duoxa1| Gene Symbol: Duoxa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK042259.1 | - | 2 | 316 | ||
| BC019755.1 | - | 8 | 781 | 3964798 | ||