siRNA Details
Gene Symbol: Prepl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163622.1 | NP_001157094.1 | 11, 12 | 2236 | ||
NM_001163623.1 | NP_001157095.1 | 10, 11 | 1700 | |||
NM_001163624.1 | NP_001157096.1 | 10, 11 | 1568 | |||
NM_145984.3 | NP_666096.3 | 11, 12 | 1800 | |||
GenBank mRNA | AK028877.1 | - | 11, 12 | 2194 | ||
AK050878.1 | - | 10, 11 | 1562 | |||
AK083070.1 | - | 11, 12 | 1741 | |||
AK129142.1 | - | 5, 6 | 1654 | |||
BC004612.1 | - | 10, 11 | 1654 | 3585402 |
