siRNA Details
| Gene Symbol: Prepl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163622.1 | NP_001157094.1 | 6 | 1123 | ||
| NM_001163623.1 | NP_001157095.1 | 5 | 587 | |||
| NM_001163624.1 | NP_001157096.1 | 5 | 455 | |||
| NM_145984.3 | NP_666096.3 | 6 | 687 | |||
| GenBank mRNA | AK028877.1 | - | 6 | 1081 | ||
| AK050878.1 | - | 5 | 449 | |||
| AK083070.1 | - | 6 | 628 | |||
| AK163108.1 | - | 6 | 1088 | |||
| BC004612.1 | - | 5 | 541 | 3585402 | ||
| BC129880.1 | - | 4 | 1119 | |||
| BY257815.1 | - | 5 | 456 | |||

Prepl
Genomic Map