siRNA Details
| Gene Symbol: Prepl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163622.1 | NP_001157094.1 | 7 | 1366 | ||
| NM_001163623.1 | NP_001157095.1 | 6 | 830 | |||
| NM_001163624.1 | NP_001157096.1 | 6 | 698 | |||
| NM_145984.3 | NP_666096.3 | 7 | 930 | |||
| GenBank mRNA | AK028877.1 | - | 7 | 1324 | ||
| AK050878.1 | - | 6 | 692 | |||
| AK083070.1 | - | 7 | 871 | |||
| AK129142.1 | - | 1 | 784 | |||
| BC004612.1 | - | 6 | 784 | 3585402 | ||

Prepl
Genomic Map