siRNA Details
| Gene Symbol: Skp1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011543.4 | NP_035673.3 | 2 | 166 | ||
| GenBank mRNA | AF083214.1 | - | 2 | 90 | ||
| AK002983.1 | - | 2 | 137 | |||
| AK010628.1 | - | 2 | 152 | |||
| AK012498.1 | - | 2 | 125 | |||
| AK014245.1 | - | 2 | 125 | |||
| AK027909.1 | - | 2 | 124 | |||
| AK078330.1 | - | 2 | 152 | |||
| AK088339.1 | - | 2 | 125 | |||
| AK152609.1 | - | 2 | 124 | |||
| AK160198.1 | - | 2 | 125 | |||
| AK166673.1 | - | 2 | 125 | |||
| BC002115.1 | - | 2 | 109 | 3490159 | ||
| BU841654.1 | - | 2 | 169 | |||
| Z47088.1 | - | 2 | 115 | |||

Skp1a
Genomic Map