siRNA Details
| Gene Symbol: Gm4787 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001038995.2 | NP_001034084.2 | 1 | 1066 | ||
| GenBank mRNA | BC141094.1 | - | 1 | 1044 | 9055706 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm4787| Gene Symbol: Gm4787 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001038995.2 | NP_001034084.2 | 1 | 1066 | ||
| GenBank mRNA | BC141094.1 | - | 1 | 1044 | 9055706 | |