siRNA Details
| Gene Symbol: Gm4788 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029977.3 | NP_001025148.3 | 8 | 1321 | ||
| NM_001160303.1 | NP_001153775.1 | 8 | 1318 | |||
| NM_001160304.1 | NP_001153776.1 | 8 | 1321 | |||
| GenBank mRNA | AJ871401.1 | - | 8 | 1250 | ||
| AJ871402.1 | - | 8 | 1250 | |||
| BC141136.1 | - | 8 | 1290 | 9055748 | ||
| BC145444.1 | - | 8 | 1290 | 9054002 | ||
| M29009.1 | - | 8 | 1315 | |||

Gm4788
Genomic Map