siRNA Details
| Gene Symbol: Tcp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290712.1 | NP_001277641.1 | 6 | 925 | ||
| NM_013686.4 | NP_038714.2 | 7 | 1011 | |||
| GenBank mRNA | AK149611.1 | - | 7 | 876 | ||
| AK149755.1 | - | 7 | 877 | |||
| AK150666.1 | - | 5 | 2436 | |||
| AK150744.1 | - | 5 | 2436 | |||
| AK151068.1 | - | 7 | 876 | |||
| AK151445.1 | - | 6 | 815 | |||
| AK151903.1 | - | 5 | 2436 | |||
| AK152633.1 | - | 5 | 2436 | |||
| AK152759.1 | - | 7 | 877 | |||
| AK153037.1 | - | 7 | 877 | |||
| AK153430.1 | - | 6 | 815 | |||
| AK165665.1 | - | 7 | 878 | |||
| AK166828.1 | - | 7 | 875 | |||
| AK166966.1 | - | 7 | 899 | |||
| AK167529.1 | - | 7 | 876 | |||
| BC003809.1 | - | 7 | 859 | 3599670 | ||
| D90344.1 | - | 7 | 826 | |||
| M12899.1 | - | 7 | 834 | |||

Tcp1
Genomic Map