siRNA Details
| Gene Symbol: Cep164 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081373.2 | NP_001074842.2 | 27 | 3539 | ||
| GenBank mRNA | AK050991.1 | - | 6 | 624 | ||
| AK220365.1 | - | 23 | 2882 | |||
| BC066145.1 | - | 9 | 981 | |||
| BC076567.1 | - | 25 | 3280 | |||

Cep164
Genomic Map