siRNA Details
| Gene Symbol: Cep164 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081373.2 | NP_001074842.2 | 3, 4 | 378 | ||
| GenBank mRNA | AK158671.1 | - | 3, 4 | 297 | ||
| BC076567.1 | - | 1, 2 | 117 | |||
| CK789347.1 | - | 3, 4 | 415 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cep164| Gene Symbol: Cep164 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081373.2 | NP_001074842.2 | 3, 4 | 378 | ||
| GenBank mRNA | AK158671.1 | - | 3, 4 | 297 | ||
| BC076567.1 | - | 1, 2 | 117 | |||
| CK789347.1 | - | 3, 4 | 415 | |||