siRNA Details
| Gene Symbol: Tdrd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134741.1 | NP_001128213.1 | 4 | 799 | ||
| NM_001277730.1 | NP_001264659.1 | 4 | 505 | |||
| GenBank mRNA | AK132674.1 | - | 4 | 785 | ||
| AK133026.1 | - | 2 | 169 | |||
| BC099972.1 | - | 4 | 505 | 30080156 | ||

Tdrd5
Genomic Map