siRNA Details
| Gene Symbol: Aldh5a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172532.3 | NP_766120.1 | 8, 9 | 1391 | ||
| GenBank mRNA | AK052703.1 | - | 8, 9 | 1392 | ||
| AK144030.1 | - | 8, 9 | 1375 | |||
| AK159485.1 | - | 8, 9 | 1371 | |||
| AK166313.1 | - | 8, 9 | 1371 | |||
| BC138703.1 | - | 8, 9 | 1354 | 8861725 | ||

Aldh5a1
Genomic Map