siRNA Details
| Gene Symbol: Pstk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039534.1 | NP_001034623.1 | 2 | 279 | ||
| GenBank mRNA | AK077585.1 | - | 2 | 280 | ||
| BC030313.1 | - | 1 | 537 | |||
| BC110381.1 | - | 2 | 255 | 5149569 | ||
| BC115503.2 | - | 2 | 280 | |||

Pstk
Genomic Map