siRNA Details
| Gene Symbol: Pstk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039534.1 | NP_001034623.1 | 5 | 841 | ||
| GenBank mRNA | AI173292.1 | - | 3 | 337 | ||
| AK077585.1 | - | 5 | 831 | |||
| BC030313.1 | - | 3 | 1333 | |||
| BC110381.1 | - | 5 | 817 | 5149569 | ||
| BC115503.2 | - | 5 | 842 | |||

Pstk
Genomic Map