siRNA Details
| Gene Symbol: Duox2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177610.2 | NP_808278.2 | 12 | 1496 | ||
| GenBank mRNA | BC172138.1 | - | 12 | 1788 | 9054808 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Duox2| Gene Symbol: Duox2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177610.2 | NP_808278.2 | 12 | 1496 | ||
| GenBank mRNA | BC172138.1 | - | 12 | 1788 | 9054808 | |